Gene Transcripts

Gene Symbol
RIC8B
Entrez Gene ID
55188
Gene Name
RIC8 guanine nucleotide exchange factor B
Gene Aliases
RIC8, hSyn
Chromosome Location
Chr. 12: 106774285 - 106889316 on Build GRCh38
UniGene ID
Hs.131306
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018157.2 NP_060627.2 3 589
XM_005268998.3 XP_005269055.1 3 615
XM_011538526.2 XP_011536828.1 2 971
XM_011538527.2 XP_011536829.1 2 877
XM_011538528.2 XP_011536830.1 2 973
XM_011538529.2 XP_011536831.1 2 968
XM_017019548.1 XP_016875037.1 3 615
XM_017019549.1 XP_016875038.1 2 480
GenBank mRNA AK001482.1 - 3 496
AY221116.1 - 3 398
BC034220.1 - 4 632 5264088
BC132689.1 - 3 570 40146711
BC144056.1 - 3 570 9052576
DA534432.1 - 1 114

View Full Product Details