Gene Transcripts

Gene Symbol
SGF29
Entrez Gene ID
112869
Gene Name
SAGA complex associated factor 29
Gene Aliases
CCDC101, STAF36, TDRD29
Chromosome Location
Chr. 16: 28553915 - 28591790 on Build GRCh38
UniGene ID
Hs.655476
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138414.2 NP_612423.1 3 285
XM_017022894.1 XP_016878383.1 3 298
GenBank mRNA AK057008.1 - 3 244
BC011981.1 - 3 276 3863665

View Full Product Details