Gene Transcripts

Gene Symbol
RMI2
Entrez Gene ID
116028
Gene Name
RecQ mediated genome instability 2
Gene Aliases
BLAP18, C16orf75
Chromosome Location
Chr. 16: 11345438 - 11351763 on Build GRCh38
UniGene ID
Hs.347524
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152308.2 NP_689521.1 2 1203
NR_130754.1 2 980
GenBank mRNA AK123764.1 - 2 1192
BC013040.2 - 2 1171 3542171
BC022427.1 - 2 1187 4252688
BC031016.1 - 2 1191 4659915
BC039361.1 - 5 1625 5270594

View Full Product Details