Gene Transcripts

Gene Symbol
NPR1
Entrez Gene ID
4881
Gene Name
natriuretic peptide receptor 1
Gene Aliases
ANPRA, ANPa, GUC2A, GUCY2A, NPRA
Chromosome Location
Chr. 1: 153678617 - 153693992 on Build GRCh38
UniGene ID
Hs.490330
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000906.3 NP_000897.3 3 1377
XM_005245218.1 XP_005245275.1 3 1448
XM_017001374.1 XP_016856863.1 3 1448
GenBank mRNA AK025024.1 - 3 1377
AK300446.1 - 2 583
BC063304.1 - 3 1426 30331757
X15357.1 - 3 999

View Full Product Details