Gene Transcripts

Gene Symbol
CEACAM8
Entrez Gene ID
1088
Gene Name
carcinoembryonic antigen related cell adhesion molecule 8
Gene Aliases
CD66b, CD67, CGM6, NCA-95
Chromosome Location
Chr. 19: 42580243 - 42595016 on Build GRCh38
UniGene ID
Hs.41
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001816.3 NP_001807.2 6 2148
GenBank mRNA BC026263.1 - 6 2112
D90064.1 - 6 2133
M33326.1 - 6 2133

View Full Product Details