Gene Transcripts

Gene Symbol
SERTAD4
Entrez Gene ID
56256
Gene Name
SERTA domain containing 4
Gene Aliases
DJ667H12.2
Chromosome Location
Chr. 1: 210232650 - 210247734 on Build GRCh38
UniGene ID
Hs.600545
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019605.3 NP_062551.1 4 1043
GenBank mRNA AK096381.1 - 4 1009
AK315386.1 - 3 1042
BC012083.1 - 4 1043 4564813

View Full Product Details