Gene Transcripts

Gene Symbol
Rab7
Entrez Gene ID
19349
Gene Name
RAB7, member RAS oncogene family
Gene Aliases
Rab7a
Chromosome Location
Chr. 6: 87999106 - 88045270 on Build GRCm38
UniGene ID
Mm.333233
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001293652.1 NP_001280581.1 5 541
NM_001293653.1 NP_001280582.1 3 202
NM_001293654.1 NP_001280583.1 4 368
NM_001293655.1 NP_001280584.1 3 281
NM_009005.3 NP_033031.2 3 319
GenBank mRNA AB232599.1 - 2 113
AK005004.1 - 3 281
AK057671.1 - 3 248
AK135659.1 - 4 369
AK139914.1 - 3 272
AK148117.1 - 3 312
AK151078.1 - 3 293
AK152666.1 - 5 516
AK154234.1 - 3 300
AK158971.1 - 3 203
AK159507.1 - 3 249
AK159928.1 - 3 220
AK179526.1 - 3 158
AK190348.1 - 3 158
AK203105.1 - 3 158
BC004597.1 - 3 237 3584891
BC086793.1 - 3 235 6414012
BY291181.1 - 3 320
CT010352.1 - 2 113
X89650.1 - 3 269

View Full Product Details