Gene Transcripts

Gene Symbol
Rab19
Entrez Gene ID
19331
Gene Name
RAB19, member RAS oncogene family
Gene Aliases
Chromosome Location
Chr. 6: 39381428 - 39390666 on Build GRCm38
UniGene ID
Mm.123866
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011226.1 NP_035356.1 4 685
GenBank mRNA AB232613.1 - 3 591
AK017590.1 - 4 718
BC032936.1 - 4 685 1379043
X80473.1 - 3 591

View Full Product Details