Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001172117.1 | NP_001165588.1 | 7 | 763 | |
NM_010407.4 | NP_034537.2 | 7 | 763 | ||
GenBank mRNA | AK149736.1 | - | 7 | 729 | |
AK150290.1 | - | 7 | 737 | ||
AK150709.1 | - | 7 | 746 | ||
AK152506.1 | - | 7 | 737 | ||
AK155975.1 | - | 7 | 676 | ||
AK159234.1 | - | 7 | 696 | ||
AK165315.1 | - | 7 | 728 | ||
BC010478.1 | - | 7 | 759 | 3978518 | |
J03023.1 | - | 7 | 652 | ||
Y00487.1 | - | 7 | 658 |