Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001159626.1 | NP_001153098.1 | 9 | 1034 | |
NM_024284.2 | NP_077246.2 | 9 | 1124 | ||
GenBank mRNA | AK147022.1 | - | 9 | 1085 | |
AK150557.1 | - | 9 | 1042 | ||
AK151231.1 | - | 9 | 1042 | ||
AK152742.1 | - | 9 | 1042 | ||
AK165777.1 | - | 9 | 1028 | ||
BC004749.1 | - | 9 | 1067 | 3583919 | |
BC019817.1 | - | 9 | 1017 | 5123648 |