Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007725.2 | NP_031751.1 | 2 | 193 | |
GenBank mRNA | AK051845.1 | - | 2 | 194 | |
AK189304.1 | - | 2 | 121 | ||
AK218463.1 | - | 1 | 55 | ||
BC008181.1 | - | 2 | 111 | 3589470 | |
BC009144.1 | - | 2 | 148 | 3594856 | |
BC018482.1 | - | 2 | 163 | 3589158 |