Gene Transcripts

Gene Symbol
Chrna6
Entrez Gene ID
11440
Gene Name
cholinergic receptor, nicotinic, alpha polypeptide 6
Gene Aliases
Acra6, Nica6
Chromosome Location
Chr. 8: 27403212 - 27413944 on Build GRCm38
UniGene ID
Mm.283137
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021369.2 NP_067344.2 5 662
GenBank mRNA AJ245706.1 - 5 500
AK149262.1 - 5 603
AY574265.1 - 5 420
BC031985.1 - 5 662 4501558

View Full Product Details