Gene Transcripts

Gene Symbol
Slc16a14
Entrez Gene ID
71781
Gene Name
solute carrier family 16 (monocarboxylic acid transporters), member 14
Gene Aliases
1110004H10Rik
Chromosome Location
Chr. 1: 84905457 - 84935281 on Build GRCm38
UniGene ID
Mm.158754
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027921.1 NP_082197.1 2, 3 499
GenBank mRNA AK049846.1 - 2, 3 551
BC023456.1 - 2, 3 499 5041164

View Full Product Details