Gene Transcripts

Gene Symbol
Klhl13
Entrez Gene ID
67455
Gene Name
kelch-like 13
Gene Aliases
1200009K10Rik, AI451128, Bklhd2, D130072F20Rik, mKIAA1309
Chromosome Location
Chr. X: 23219271 - 23365107 on Build GRCm38
UniGene ID
Mm.224306
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290476.1 NP_001277405.1 5 1051
NM_026167.4 NP_080443.1 7 1070
GenBank mRNA AK004677.1 - 7 1042
AK122491.1 - 5 1051
BC100393.1 - 6 1035 30737426

View Full Product Details