Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_197985.3 | NP_932102.2 | 3 | 380 | |
GenBank mRNA | AK039667.1 | - | 3 | 588 | |
AK046591.1 | - | 3 | 387 | ||
AK134787.1 | - | 3 | 396 | ||
AK148295.1 | - | 3 | 522 | ||
AK160534.1 | - | 3 | 397 | ||
AK161404.1 | - | 3 | 345 | ||
AY424291.1 | - | 3 | 380 | ||
BC024094.1 | - | 3 | 380 | 5132696 | |
BC064109.1 | - | 3 | 378 | 3371697 |