Gene Transcripts
- Gene Symbol
- Myo18a
- Entrez Gene ID
- 360013
- Gene Name
- myosin XVIIIA
- Gene Aliases
- MAJN, MyoPDZ, MysPDZ
- Chromosome Location
- Chr. 11: 77763251 - 77865988 on Build GRCm38
- UniGene ID
- Mm.341248
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001291212.1 |
NP_001278141.1
|
4, 5 |
1319 |
|
NM_001291213.1 |
NP_001278142.1
|
4, 5 |
1427 |
|
NM_001291214.1 |
NP_001278143.1
|
3, 4 |
286 |
|
NM_001291215.1 |
NP_001278144.1
|
4, 5 |
322 |
|
NM_011586.3 |
NP_035716.1
|
3, 4 |
1283 |
|
GenBank mRNA |
AB026497.1 |
- |
3, 4 |
1253 |
|
AK137574.1 |
- |
3, 4 |
274 |
|
AK171342.1 |
- |
3, 4 |
283 |
|
BB851963.1 |
- |
3, 4 |
288 |
|
BC138366.1 |
- |
4, 5 |
1261 |
8861386 |
BC145257.1 |
- |
4, 5 |
1369 |
|
View Full Product Details