Gene Transcripts
- Gene Symbol
- Ofcc1
- Entrez Gene ID
- 218165
- Gene Name
- orofacial cleft 1 candidate 1
- Gene Aliases
- opo
- Chromosome Location
- Chr. 13: 40001834 - 40361259 on Build GRCm38
- UniGene ID
- Mm.364096
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF548110.1 |
- |
4 |
763 |
|
AF548111.1 |
- |
3 |
499 |
|
View Full Product Details