Gene Transcripts

Gene Symbol
Dnah5
Entrez Gene ID
110082
Gene Name
dynein, axonemal, heavy chain 5
Gene Aliases
AU022615, Dnahc5, Mdnah5, b2b1134Clo, b2b1154Clo, b2b1537Clo, b2b1565Clo, b2b601Clo, mKIAA1603
Chromosome Location
Chr. 15: 28154854 - 28472045 on Build GRCm38
UniGene ID
Mm.248464
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133365.3 NP_579943.3 3 431
GenBank mRNA AF466704.1 - 3 432
AK054152.1 - 3 433

View Full Product Details