Gene Transcripts

Gene Symbol
Sgms1
Entrez Gene ID
208449
Gene Name
sphingomyelin synthase 1
Gene Aliases
9530058O11Rik, AI841905, C80702, Mob, Sms1, Sor1, Tmem23
Chromosome Location
Chr. 19: 32122727 - 32389705 on Build GRCm38
UniGene ID
Mm.329810
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001168525.1 NP_001161997.1 8 1947
NM_001168526.1 NP_001161998.1 4 1171
NM_144792.4 NP_659041.3 8 1515
GenBank mRNA AK076554.1 - 8 1315
AK079254.1 - 4 1116
AK133967.1 - 4 1139
AK146782.1 - 8 1515
AK150272.1 - 4 1148
AK151170.1 - 8 1500
AK152312.1 - 4 1149
AK153406.1 - 8 1500
AK160539.1 - 4 1149
AY509044.1 - 8 1515
BC019443.1 - 4 1138 5041121
BC076618.1 - 8 1947
BC085298.1 - 5 1109 6307174

View Full Product Details