Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001271725.1 | NP_001258654.1 | 12 | 4082 | |
NM_001271726.1 | NP_001258655.1 | 12 | 4118 | ||
NM_001271727.1 | NP_001258656.1 | 12 | 4387 | ||
NM_001271728.1 | NP_001258657.1 | 12 | 4185 | ||
NM_030706.3 | NP_109631.2 | 12 | 4181 | ||
GenBank mRNA | AB043550.1 | - | 12 | 4159 | |
AK054540.1 | - | 1 | 584 | ||
AK147275.1 | - | 12 | 4295 | ||
AK147395.1 | - | 12 | 4176 | ||
AK164350.1 | - | 11 | 3884 | ||
BC058961.1 | - | 12 | 4185 | 6826143 |