Gene Transcripts

Gene Symbol
Tbl1xr1
Entrez Gene ID
81004
Gene Name
transducin (beta)-like 1X-linked receptor 1
Gene Aliases
8030499H02Rik, A630076E03Rik, AW539987, C21, C230089I12Rik, DC42, Ira1, TBLR1
Chromosome Location
Chr. 3: 22076593 - 22216594 on Build GRCm38
UniGene ID
Mm.202966
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030732.3 NP_109657.2 16 2468
GenBank mRNA AK033347.1 - 16 2338
AK136255.1 - 16 2290
BC018512.1 - 6 1114

View Full Product Details