Gene Transcripts
- Gene Symbol
- Zc3h18
- Entrez Gene ID
- 76014
- Gene Name
- zinc finger CCCH-type containing 18
- Gene Aliases
- 1190001B23Rik, 5830416A07Rik, Nhn1
- Chromosome Location
- Chr. 8: 122376614 - 122417360 on Build GRCm38
- UniGene ID
- Mm.241489
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001029993.1 |
NP_001025164.1
|
17, 18 |
2830 |
|
NM_001029994.1 |
NP_001025165.1
|
16, 17 |
2758 |
|
GenBank mRNA |
AK031413.1 |
- |
17, 18 |
2780 |
|
AK131189.1 |
- |
14, 15 |
1902 |
|
AK159881.1 |
- |
16, 17 |
2700 |
|
AK165989.1 |
- |
15, 16 |
2113 |
|
AK169952.1 |
- |
6, 7 |
3819 |
|
BC003319.1 |
- |
7, 8 |
1002 |
|
BC030495.1 |
- |
16, 17 |
2738 |
5400494 |
BC094330.1 |
- |
14, 15 |
1911 |
|
View Full Product Details