Gene Transcripts
- Gene Symbol
- Ier5l
- Entrez Gene ID
- 72500
- Gene Name
- immediate early response 5-like
- Gene Aliases
- 2610524G09Rik
- Chromosome Location
- Chr. 2: 30472631 - 30474199 on Build GRCm38
- UniGene ID
- Mm.295464
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK034055.1 |
- |
|
2462 |
|
View Full Product Details