Gene Transcripts

Gene Symbol
Ncapg2
Entrez Gene ID
76044
Gene Name
non-SMC condensin II complex, subunit G2
Gene Aliases
5830426I05Rik, Luzp5, Mtb, mCAP-G2
Chromosome Location
Chr. 12: 116405357 - 116463532 on Build GRCm38
UniGene ID
Mm.491196
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133762.3 NP_598523.3 15 1934
GenBank mRNA AY455829.1 - 9 1045
BC008539.1 - 4 615
BC076631.1 - 14 1744 30637859

View Full Product Details