Gene Transcripts

Gene Symbol
Herc4
Entrez Gene ID
67345
Gene Name
hect domain and RLD 4
Gene Aliases
1700056O17Rik, 4921531D01Rik, 9530080M15Rik, mKIAA1593
Chromosome Location
Chr. 10: 63243782 - 63317881 on Build GRCm38
UniGene ID
Mm.234437
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026101.4 NP_080377.2 23 3275
NM_030114.2 NP_084390.1 24 3299
GenBank mRNA AK019546.1 - 21 2907
AK143111.1 - 24 3173
AK145735.1 - 18 2223
AK147723.1 - 23 3162
BC026855.1 - 9 1032
BC043082.1 - 23 3087 6417111
BC047157.1 - 9 1177
BC060033.1 - 22 2884 6478123

View Full Product Details