Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_016876.3 | NP_058572.2 | 6, 7 | 440 | |
GenBank mRNA | AF108214.1 | - | 6, 7 | 454 | |
AK038249.1 | - | 6, 7 | 450 | ||
AK168362.1 | - | 6, 7 | 443 | ||
BC008511.1 | - | 6, 7 | 440 | 3592537 | |
U70733.1 | - | 6, 7 | 430 |