Gene Transcripts

Gene Symbol
Tgoln1
Entrez Gene ID
22134
Gene Name
trans-golgi network protein
Gene Aliases
AU015504, D6Ertd384e, TGN38, TGN38A, Ttgn1
Chromosome Location
Chr. 6: 72608420 - 72617000 on Build GRCm38
UniGene ID
Mm.246563
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009443.3 NP_033469.1 4 2936
GenBank mRNA AK076586.1 - 4 2937
AK133559.1 - 4 2683
AK161242.1 - 5 2696

View Full Product Details