Gene Transcripts
- Gene Symbol
- Tbrg4
- Entrez Gene ID
- 21379
- Gene Name
- transforming growth factor beta regulated gene 4
- Gene Aliases
- 2310042P22Rik, AA120735, AA408001, AI527316, Cpr2, R74877, Tb-12, mKIAA0948
- Chromosome Location
- Chr. 11: 6615598 - 6626084 on Build GRCm38
- UniGene ID
- Mm.247761
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001130457.1 |
NP_001123929.1
|
3, 4 |
629 |
|
NM_134011.2 |
NP_598772.1
|
3, 4 |
629 |
|
GenBank mRNA |
AK028966.1 |
- |
3, 4 |
607 |
|
AK144813.1 |
- |
3, 4 |
627 |
|
AK166760.1 |
- |
4, 5 |
537 |
|
AK168088.1 |
- |
3, 4 |
627 |
|
AK188017.1 |
- |
1, 2 |
244 |
|
BC016483.1 |
- |
3, 4 |
585 |
3155271 |
BC031155.1 |
- |
3, 4 |
600 |
4976690 |
BC031910.1 |
- |
3, 4 |
599 |
5362943 |
View Full Product Details