Gene Transcripts
- Gene Symbol
- PWP1
- Entrez Gene ID
- 11137
- Gene Name
- PWP1 homolog, endonuclein
- Gene Aliases
- IEF-SSP-9502
- Chromosome Location
- Chr. 12: 107685732 - 107713162 on Build GRCh38
- UniGene ID
- Hs.506652
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001317962.1 |
NP_001304891.1
|
1, 2 |
104 |
|
NM_001317963.1 |
NP_001304892.1
|
1, 2 |
248 |
|
NM_007062.2 |
NP_008993.1
|
1, 2 |
248 |
|
GenBank mRNA |
AK223162.1 |
- |
1, 2 |
172 |
|
AK290681.1 |
- |
1, 2 |
89 |
|
AK296252.1 |
- |
1, 2 |
105 |
|
AK297961.1 |
- |
1, 2 |
200 |
|
BC000067.2 |
- |
1, 2 |
131 |
|
BC001652.2 |
- |
1, 2 |
167 |
2924215 |
BC010921.1 |
- |
1, 2 |
182 |
|
BC032127.2 |
- |
1, 2 |
170 |
|
BC040135.1 |
- |
1, 2 |
146 |
|
BC046144.1 |
- |
1, 2 |
162 |
|
DC380488.1 |
- |
1, 2 |
248 |
|
L07758.1 |
- |
1, 2 |
167 |
|
View Full Product Details