Gene Transcripts

Gene Symbol
Siah1
Entrez Gene ID
140941
Gene Name
siah E3 ubiquitin protein ligase 1
Gene Aliases
SIAH, Siah1a
Chromosome Location
Chr. 19: 21372163 - 21395773 on Build Rnor_6.0
UniGene ID
Rn.73937
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080905.2 NP_543181.2 2 818
XM_006255211.2 XP_006255273.1 2 5419
XM_006255213.2 XP_006255275.1 2 1158
XM_008772345.1 XP_008770567.1 3 859
GenBank mRNA AB067814.1 - 2 677
AF389476.1 - 1 638
AY681965.1 - 1 638

View Full Product Details