Gene Transcripts

Gene Symbol
Brs3
Entrez Gene ID
260319
Gene Name
bombesin-like receptor 3
Gene Aliases
Chromosome Location
Chr. X: 159484954 - 159510944 on Build Rnor_6.0
UniGene ID
Rn.86415
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152845.1 NP_690058.1 1 215
XM_006257677.2 XP_006257739.1 2 245
XM_006257678.2 XP_006257740.1 2 64
GenBank mRNA AF510984.1 - 1 215

View Full Product Details