Gene Transcripts

Gene Symbol
Tubg1
Entrez Gene ID
252921
Gene Name
tubulin, gamma 1
Gene Aliases
Chromosome Location
Chr. 10: 89030865 - 89037455 on Build Rnor_6.0
UniGene ID
Rn.154431
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145778.2 NP_665721.1 5, 6 587
GenBank mRNA AB015946.1 - 5, 6 492
BC070957.1 - 5, 6 587 7105643

View Full Product Details