Gene Transcripts

Gene Symbol
Ctgf
Entrez Gene ID
64032
Gene Name
connective tissue growth factor
Gene Aliases
CTGRP
Chromosome Location
Chr. 1: 21851657 - 21854773 on Build Rnor_6.0
UniGene ID
Rn.17145
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022266.2 NP_071602.1 5 1290
GenBank mRNA AB023068.1 - 5 1278
AF120275.1 - 5 1290
BC072503.1 - 4 1659 7098774

View Full Product Details