Gene Transcripts

Gene Symbol
FBXO2
Entrez Gene ID
26232
Gene Name
F-box protein 2
Gene Aliases
FBG1, FBX2, Fbs1, NFB42, OCP1
Chromosome Location
Chr. 1: 11648361 - 11654831 on Build GRCh38
UniGene ID
Hs.132753
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012168.5 NP_036300.2 6 1440
GenBank mRNA AF187318.1 - 6 1288
BC025233.1 - 6 1024 5090925
BC096747.1 - 6 1040 5259298

View Full Product Details