Gene Transcripts

Gene Symbol
Lrrc15
Entrez Gene ID
246296
Gene Name
leucine rich repeat containing 15
Gene Aliases
Lib
Chromosome Location
Chr. 11: 74041102 - 74052128 on Build Rnor_6.0
UniGene ID
Rn.213072
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145083.1 NP_659551.1 1 521
XM_006248482.2 XP_006248544.1 2 637
GenBank mRNA AB071036.1 - 1 521

View Full Product Details