Gene Transcripts

Gene Symbol
Sgms1
Entrez Gene ID
353229
Gene Name
sphingomyelin synthase 1
Gene Aliases
Mob, Tmem23
Chromosome Location
Chr. 1: 250692448 - 250951685 on Build Rnor_6.0
UniGene ID
Rn.55975
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181386.2 NP_852051.2 6 920
GenBank mRNA AY280961.1 - 2 503
BC078690.1 - 1 187 7113038
BC081693.1 - 6 920 7132545

View Full Product Details