Gene Transcripts

Gene Symbol
Impdh2
Entrez Gene ID
301005
Gene Name
IMP (inosine 5'-monophosphate) dehydrogenase 2
Gene Aliases
IMPD 2, IMPDH 2
Chromosome Location
Chr. 8: 117346988 - 117351619 on Build Rnor_6.0
UniGene ID
Rn.8093
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199099.2 NP_954530.1 5 431
XM_006243751.2 XP_006243813.1 5 472
GenBank mRNA BC060585.1 - 5 432

View Full Product Details