Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001142522.1 | NP_001135994.1 | 2 | 552 | |
NM_012177.3 | NP_036309.1 | 2 | 392 | ||
GenBank mRNA | AF129535.1 | - | 2 | 344 | |
AJ420499.1 | - | 2 | 412 | ||
AK026197.1 | - | 3 | 301 | ||
AK055221.1 | - | 2 | 526 | ||
AK314456.1 | - | 2 | 374 | ||
AY079515.1 | - | 2 | 283 | ||
BC018905.1 | - | 2 | 334 | 3960052 | |
BG944882.1 | - | 2 | 364 | ||
BI838832.1 | - | 2 | 583 | ||
DB122061.1 | - | 2 | 392 |