Gene Transcripts

Gene Symbol
Acpp
Entrez Gene ID
56780
Gene Name
acid phosphatase, prostate
Gene Aliases
5'-NT, Acpp11, FRAP, Ppal, RNACPP11, pap
Chromosome Location
Chr. 8: 112838629 - 112884062 on Build Rnor_6.0
UniGene ID
Rn.40121
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134901.1 NP_001128373.1 2 201
NM_020072.1 NP_064457.1 2 241
GenBank mRNA DQ826426.1 - 2 201
M32397.1 - 2 241

View Full Product Details