Gene Transcripts

Gene Symbol
FBXL7
Entrez Gene ID
23194
Gene Name
F-box and leucine rich repeat protein 7
Gene Aliases
FBL6, FBL7
Chromosome Location
Chr. 5: 15500196 - 15939796 on Build GRCh38
UniGene ID
Hs.433057
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278317.1 NP_001265246.1 4 3663
NM_012304.4 NP_036436.1 4 3895
XM_011513998.1 XP_011512300.1 4 3454
XM_017009262.1 XP_016864751.1 4 7875
GenBank mRNA AB020647.2 - 4 3895
AK021671.1 - 1 863
BC050571.1 - 1 1480

View Full Product Details