Gene Transcripts

Gene Symbol
Rdh16
Entrez Gene ID
299511
Gene Name
retinol dehydrogenase 16 (all-trans)
Gene Aliases
Rdh2, RoDHII
Chromosome Location
Chr. : - on Build
UniGene ID
Rn.94108
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199208.1 NP_954678.1 1003
GenBank mRNA BC062000.1 - 970
BC079153.1 - 4 827 7128686
U33500.1 - 1003

View Full Product Details