Gene Transcripts

Gene Symbol
FLRT1
Entrez Gene ID
23769
Gene Name
fibronectin leucine rich transmembrane protein 1
Gene Aliases
SPG68
Chromosome Location
Chr. 11: 64035970 - 64119183 on Build GRCh38
UniGene ID
Hs.584876
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013280.4 NP_037412.2 2 676
XM_005273861.1 XP_005273918.1 3 1562
XM_011544883.1 XP_011543185.1 2 516
GenBank mRNA AF169675.1 - 1 335
AK289931.1 - 2 993
AY358308.1 - 2 797
BC018370.1 - 2 676 4214494

View Full Product Details