Gene Transcripts

Gene Symbol
AKAP8L
Entrez Gene ID
26993
Gene Name
A-kinase anchoring protein 8 like
Gene Aliases
HA95, HAP95, NAKAP, NAKAP95
Chromosome Location
Chr. 19: 15380048 - 15419121 on Build GRCh38
UniGene ID
Hs.399800
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291478.1 NP_001278407.1 4 365
NM_014371.3 NP_055186.3 4 365
NR_111971.1 4 365
XM_005259854.3 XP_005259911.2 4 272
XM_011527916.2 XP_011526218.1 4 272
GenBank mRNA AB015332.1 - 4 212
AB025905.1 - 4 212
AB451326.1 - 3 167
AB451469.1 - 3 167
AF199414.1 - 3 167
AJ243467.1 - 4 213
AK001594.1 - 4 232
AK094736.1 - 4 232
AK225046.1 - 4 232
AK225291.1 - 4 232
AK295956.1 - 4 270
BC000713.2 - 4 217 3504388
CD244113.1 - 4 216
DA315789.1 - 4 349
DB099586.1 - 4 365

View Full Product Details