Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001282873.1 | NP_001269802.1 | 12 | 1876 | |
NM_015902.5 | NP_056986.2 | 12 | 1876 | ||
XM_011517104.2 | XP_011515406.2 | 11 | 1861 | ||
XM_011517105.2 | XP_011515407.2 | 11 | 1861 | ||
XM_011517106.2 | XP_011515408.2 | 11 | 1843 | ||
XM_017013533.1 | XP_016869022.1 | 11 | 1861 | ||
XM_017013534.1 | XP_016869023.1 | 11 | 1861 | ||
GenBank mRNA | AB020703.3 | - | 12 | 1876 | |
AF006010.2 | - | 12 | 1454 | ||
BC137234.1 | - | 12 | 1448 | 9021231 | |
U95000.1 | - | 12 | 1418 |