Gene Transcripts

Gene Symbol
SLC39A2
Entrez Gene ID
29986
Gene Name
solute carrier family 39 member 2
Gene Aliases
6A1, ETI-1, ZIP-2, ZIP2
Chromosome Location
Chr. 14: 20999255 - 21001875 on Build GRCh38
UniGene ID
Hs.175783
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256588.1 NP_001243517.1 2 378
NM_014579.3 NP_055394.2 2 378
GenBank mRNA AF186081.1 - 2 322
AK314974.1 - 2 342
BC096723.1 - 2 209 40004409
BC098254.1 - 2 209 40004407
BC110057.1 - 2 209 40004405
BP359281.1 - 2 380
CN306082.1 - 2 304
D83039.1 - 2 316
DA871745.1 - 2 342

View Full Product Details