Gene Transcripts
- Gene Symbol
- GANAB
- Entrez Gene ID
- 23193
- Gene Name
- glucosidase II alpha subunit
- Gene Aliases
- G2AN, GIIA, GLUII
- Chromosome Location
- Chr. 11: 62624826 - 62646726 on Build GRCh38
- UniGene ID
- Hs.595071
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_198334.2 |
NP_938148.1
|
3, 4 |
387 |
|
NM_198335.3 |
NP_938149.2
|
3, 4 |
387 |
|
XM_017017412.1 |
XP_016872901.1
|
1, 2 |
121 |
|
XM_017017413.1 |
XP_016872902.1
|
3, 4 |
450 |
|
XM_017017414.1 |
XP_016872903.1
|
3, 4 |
281 |
|
GenBank mRNA |
AF144074.1 |
- |
3, 4 |
276 |
|
AJ000332.1 |
- |
3, 4 |
260 |
|
AK026997.1 |
- |
3, 4 |
277 |
|
AK314370.1 |
- |
3, 4 |
276 |
|
BG720650.1 |
- |
3, 4 |
332 |
|
D42041.1 |
- |
3, 4 |
258 |
|
DC400833.1 |
- |
3, 4 |
387 |
|
EU794613.1 |
- |
3, 4 |
293 |
|
View Full Product Details