Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_003959.2 | NP_003950.1 | 27 | 2691 | |
XM_011538963.1 | XP_011537265.1 | 27 | 2994 | ||
GenBank mRNA | AB013384.1 | - | 21 | 2044 | |
AB014555.1 | - | 27 | 2625 | ||
AK024062.1 | - | 5 | 281 | ||
AK295389.1 | - | 5 | 227 | ||
AK307446.1 | - | 4 | 268 | ||
AK308538.1 | - | 5 | 372 |