Gene Transcripts

Gene Symbol
OR2L3
Entrez Gene ID
391192
Gene Name
olfactory receptor family 2 subfamily L member 3
Gene Aliases
Chromosome Location
Chr. 1: 248060682 - 248061620 on Build GRCh38
UniGene ID
Hs.690412
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004687.1 NP_001004687.1 1 43
GenBank mRNA BC137039.1 - 1 63
BC137040.1 - 1 63 9021036

View Full Product Details