Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001007714.1 | NP_001007715.1 | 4 | 434 | |
XM_006257295.1 | XP_006257357.1 | 4 | 470 | ||
XM_006257296.2 | XP_006257358.1 | 4 | 464 | ||
XM_006257297.1 | XP_006257359.1 | 3 | 369 | ||
GenBank mRNA | AY539870.1 | - | 1 | 149 | |
BC083606.1 | - | 4 | 434 | 7133330 | |
FQ212552.1 | - | 3 | 332 | ||
FQ232538.1 | - | 4 | 433 |