Gene Transcripts

Gene Symbol
Enoph1
Entrez Gene ID
305177
Gene Name
enolase-phosphatase 1
Gene Aliases
RGD1309016
Chromosome Location
Chr. 14: 11172359 - 11198273 on Build Rnor_6.0
UniGene ID
Rn.98105
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009391.1 NP_001009391.1 5 705
XM_008770023.1 XP_008768245.1 5 755
GenBank mRNA BC087697.1 - 5 705

View Full Product Details